Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylchlorodimethylsilane, 98%

Catalog No. p-3213203
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle

CAS: 18162-48-6 | C6H15ClSi | 150.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18162-48-6
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.73
MDL Number MFCD00000501
InChI Key BCNZYOJHNLTNEZ-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
PubChem CID 28928
ChEBI CHEBI:85071
IUPAC Name tert-butyl-chloro-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)Cl

Specifications

Melting Point 85.0°C to 91.0°C
Color Colorless to White
Density 0.8700g/mL
Boiling Point 125.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)3CSi(CH3)2Cl
Quantity 5 g
UN Number 2925
Fieser 04,176; 05,74; 06,78; 08,59; 09,77; 10,62; 13,87; 17,5
Specific Gravity 0.87
Merck Index 15, 1567
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohols, organic acids and amines, soluble in ethers and hydrocarbons
Formula Weight 150.73
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material tert-Butylchlorodimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha
EINECSNumber 242-042-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.