Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyldimethylsilyl chloride, 50 wt.% solution in toluene, AcroSeal™

Catalog No. ac433111000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 18162-48-6 | C6H15ClSi | 150.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyldimethylsilyl chloride
CAS 18162-48-6, 108-88-3
Color Colorless
Density 0.8660g/mL
Boiling Point 110.0°C to 126.0°C
Flash Point 4°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C6H15ClSi
Quantity 100 mL
Specific Gravity 0.866
Merck Index 15,1567
Synonym tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
InChI Key BCNZYOJHNLTNEZ-UHFFFAOYSA-N
SMILES CC(C)(C)[Si](C)(C)Cl
IUPAC Name tert-butyl-chloro-dimethylsilane
Molecular Weight (g/mol) 150.72
PubChem CID 28928
ChEBI CHEBI:85071
Formula Weight 150.72
Percent Purity 48 to 52%
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.