Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylferrocene, 98%, Thermo Scientific Chemicals

Catalog No. p-7050756
Click to view available options
Quantity:
1 g
5 g

It finds its application in microwave-enhanced electrochemical processes in micellar surfactant media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1316-98-9
MDL Number MFCD00058729

Specifications

Density 1.201 g/mL
Boiling Point 96°C (1 mmHg)
Refractive Index 1.579
Quantity 1 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 242.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material tert-Butylferrocene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.