Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butylhydroquinone, 97%
Click to view available options
Quantity:
500 g
1000 g
2500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
TBHQ was used to study the inactivation of barotolerant strains of Listeria monocytogenes and Escherichia coli. Environment friendly electrode materials for supercapacitors were attained by decorating the surface of graphene nanosheets with TBHQ. TBHQ is frequently used as a food preservative. Inhibition of oxidative rancidity in frozen cooked fish flakes by tert-butylhydroquinone. Nuclear factor E2-related factor 2-dependent antioxidant response element activation is by tert-butylhydroquinoneoccurring preferentially in Astrocytes conditions.
Solubility
Slightly soluble in water(10g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TBHQ was used to study the inactivation of barotolerant strains of Listeria monocytogenes and Escherichia coli. Environment friendly electrode materials for supercapacitors were attained by decorating the surface of graphene nanosheets with TBHQ. TBHQ is frequently used as a food preservative. Inhibition of oxidative rancidity in frozen cooked fish flakes by tert-butylhydroquinone. Nuclear factor E2-related factor 2-dependent antioxidant response element activation is by tert-butylhydroquinoneoccurring preferentially in Astrocytes conditions.
Solubility
Slightly soluble in water(10g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 126°C to 129°C |
| Density | 1.05 |
| Boiling Point | 295°C |
| Flash Point | 171°C (339°F) |
| Quantity | 2500 g |
| UN Number | UN3077 |
| Beilstein | 637923 |
| Solubility Information | Slightly soluble in water(10g/L). |
| Formula Weight | 166.22 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H302+H312-H315-H317-H319-H335-H341-H361 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 217-752-2 |
| RTECSNumber | MX4375000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction