Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Nonyl Mercaptan (mixture) 97.0+%, TCI America™
SDP

Catalog No. N029925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N029925ML 25 mL
N0299500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N029925ML Supplier TCI America Supplier No. N029925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25360-10-5
Molecular Formula C9H20S
Molecular Weight (g/mol) 160.32
MDL Number MFCD00059963
InChI Key MPBLPZLNKKGCGP-UHFFFAOYSA-N
PubChem CID 520196
IUPAC Name 2-methyloctane-2-thiol
SMILES CCCCCCC(C)(C)S

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 160.32
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material tert-Nonyl Mercaptan (mixture)
EINECSNumber (2)-0464
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.