Learn More
Terumo™ SurGuard™3 Safety Hypodermic Needles, 20G
Catalog No. 22272103 Shop All Terumo Medical Corporation Products
Click to view available options
Length (English) Needle:
1 in.
1.5 in.
2 in.
This item is not returnable. View return policy
This item is not returnable. View return policy
With three ways to activate (finger, thumb and hard surface) and a broad range of sizes, SurGuard™3 is the industry's most adaptable safety needle.
  • 3 modes of activation-thumb, finger, or hard surface
  • Separate finger/thumb grips with ridges and nonslip surfaces
  • Hub and needle locks for security post activation
  • Broad range of sizes
  • Firm injection feel
  • Sharpest needles among major brands tested; 10% sharper than similar needles from Becton, Dickinson and Company
  • More comfortable injections for patients
Specifications
Needle Gauge 20 G
Latex Free Yes
Length (English) Needle 1.5 in.
Product Type Hypodermic Safety Needle
Needle Point Style Sharp
Sterility Sterile
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.