Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TES, 0.2 M Buffer Solution, pH 7.5, PROMO, Thermo Scientific Chemicals

Catalog No. J61350AEPM
Click to view available options
Quantity:
100 mL

Applications

TES, 0.2M buffer solution, is a suitable buffer for epidermal cell growth at pH 7.4 to 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.

Solubility

Soluble in water (350 g/L) at 20°C, and methanol (very faint turbidity).

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70331-82-7
Molecular Formula C6H14NNaO6S
Molecular Weight (g/mol) 251.23 g/mol
MDL Number MFCD00065482
InChI Key GUXMFAHOYNRHBI-UHFFFAOYSA-M
PubChem CID 23702152
IUPAC Name sodium;2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonate
SMILES [Na+].OCC(CO)(CO)NCCS([O-])(=O)=O

Specifications

Concentration 0.2 M
pH 7.5
Physical Form Liquid
Quantity 100 mL
Solubility Information Soluble in water at 20°C, and methanol.
Chemical Name or Material TES buffer solution

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.