Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TES, Free Acid, G-Biosciences

Click to view available options
Quantity:
500 g
1000 g
Description
It has also been used in buffers for cell culture medium.
Specifications
Specifications
Quantity | 1000 g |
Form | Powder |
Percent Purity | >95% |
Grade | Reagent |
Chemical Name or Material | TES |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction