Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TES Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553731
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553731 Supplier Invitrogen™ Supplier No. PA553731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YCDSEKPRCA GCDELIFSNE YTQAENQNWH LKHFCCFDCD SILAGEIYVM VNDKPVCKPC YVKNHAVVCQ GCHNAIDPEV QRVTYNNFSW HASTECFLCS CCSKCLIGQK FMPVEGMVFC SVECKK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

TES maps to a common fragile site on chromosome 7q31.2 designated FRA7G and this gene is similar to mouse Testin, a testosterone-responsive gene encoding a Sertoli cell secretory protein containing three LIM domains. LIM domains are double zinc-finger motifs that mediate protein-protein interactions between transcription factors, cytoskeletal proteins and signaling proteins. Multiple protein isoforms are encoded by transcript variants of the TES gene and Cancer-associated chromosomal changes often involve regions containing fragile sites involving the TES gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TES
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TES
Gene Accession No. Q9UGI8
Gene Alias D6Ertd352e; TES; Tes1; TES1/TES2; Tes2; TESS; TESS-2; testin; testin 2; testin LIM domain protein; testin2; testis derived transcript; testis derived transcript (3 LIM domains)
Gene Symbols TES
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TES. Recombinant protein control fragment (Product #RP-90902).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26136
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.