Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TESK2 Monoclonal Antibody (5H4)
TESK2 Monoclonal Antibody (5H4)
Supplier: Thermo Scientific MA521035
Description
TESK2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA), ELISA

Specifications
TESK2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q96S53 | |
TESK2 | |
TESK2 (AAH33085, 405 a.a. ~ 542 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
5H4 | |
Unconjugated | |
TESK2 | |
dual specificity testis-specific protein kinase 2; TESK2; Testicular protein kinase 2; testis-specific kinase 2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10420 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction