Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TESK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111574 Supplier Invitrogen™ Supplier No. PA5111574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FDLDAPGPGT MPLADWQEPL APPIRRWRSL PGSPEFLHQE ACPFVGREES LSDGPPPRLS SLKYRVKEIP PF Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 89%.

TESK2 or testis-specific kinase 2 is a serine/threonine protein kinase that contains an N-terminal protein kinase domain and belongs to the TESK subgroup of the LIMK/TESK family of protein kinases. The kinase domain of TESK2 is structurally similar to the kinase domain of testis-specific protein kinase-1 and the LIM motif-containing protein kinases (LIMKs). TESK2 is predominantly expressed in testis and prostate and plays an important role in spermatogenesis. TESK2 phosphorylates cofilin specifically at Ser-3 and induces formation of actin stress fibers and focal adhesions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TESK2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TESK2
Gene Accession No. Q96S53
Gene Alias dual specificity testis-specific protein kinase 2; TESK2; Testicular protein kinase 2; testis-specific kinase 2
Gene Symbols TESK2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TESK2. Recombinant protein control fragment (Product #RP-107293).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10420
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.