Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TESK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596687 Supplier Invitrogen™ Supplier No. PA596687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FVEIGKTLEE ILSRLQEEEQ ERDRKLQPTA RGLLEKAPGV KRLSSLDDKI PHKSPCPRRT IWLSRSQSDI FSRKPPRTVS VLDPYYRPRD GAARTPKVNP FSARQDLMGG KIKFFDLPSK SVISLVFDLD APGPGTMPLA DWQEPLAPPI RRWRSLPGSP EFLHQEACPF VGREESLSDG PPPRLSSLKY RVKEIPPFRA SALPAAQAHE AMDCSILQEE NGFGSRPQGT SPCPAGASEE MEVEERPAGS TPATFSTSGI GLQTQGKQDG; Positive Samples: Mouse liver, Mouse kidney; Cellular Location: Nucleus.

TESK2 or testis-specific kinase 2 is a serine/threonine protein kinase that contains an N-terminal protein kinase domain and belongs to the TESK subgroup of the LIMK/TESK family of protein kinases. The kinase domain of TESK2 is structurally similar to the kinase domain of testis-specific protein kinase-1 and the LIM motif-containing protein kinases (LIMKs). TESK2 is predominantly expressed in testis and prostate and plays an important role in spermatogenesis. TESK2 phosphorylates cofilin specifically at Ser-3 and induces formation of actin stress fibers and focal adhesions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TESK2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TESK2
Gene Accession No. Q8VCT9, Q924U5
Gene Alias dual specificity testis-specific protein kinase 2; TESK2; Testicular protein kinase 2; testis-specific kinase 2
Gene Symbols TESK2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 302-571 of human TESK2 (NP_0091012).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170908, 230661
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.