Learn More
Tesmin Monoclonal Antibody (4H4)

Description
Tesmin Monoclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
Specifications
| Antigen | Tesmin |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4H4 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TESMIN |
| Gene Accession No. | Q9Y4I5 |
| Gene Alias | CXC domain containing 2; CXCDC2; Metallothionein-like 5, testis-specific; metallothionein-like 5, testis-specific (tesmin); MTL5; MTLT; tesmin; testis expressed metallothionein like protein; testis-specific metallothionein-like protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.