Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tesmin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544081 Supplier Invitrogen™ Supplier No. PA544081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EAYLGPADPK EPVLHAFNPA LGADCKGQVK AKLAGGDSDG GELLGEYPGI Sequence homology: Dog: 83%; Human: 100%.

Metallothionein proteins are highly conserved low-molecular-weight cysteine-rich proteins that are induced by and bind to heavy metal ions and have no enzymatic activity. They may play a central role in the regulation of cell growth and differentiation and are involved in spermatogenesis. This gene encodes a metallothionein-like protein which has been shown to be expressed differentially in mouse testis and ovary. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tesmin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TESMIN
Gene Accession No. Q9Y4I5
Gene Alias CXC domain containing 2; CXCDC2; Metallothionein-like 5, testis-specific; metallothionein-like 5, testis-specific (tesmin); MTL5; MTLT; tesmin; testis expressed metallothionein like protein; testis-specific metallothionein-like protein
Gene Symbols TESMIN
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MTL5 (aa 51-100).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9633
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.