Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TESPA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546497 Supplier Invitrogen™ Supplier No. PA546497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SLPIEDPQWS TDPAQIRREL CSLPATNTET HPAKDETFWK RKSRARKSLF Sequence homology: Dog: 93%; Horse: 100%; Human: 100%; Pig: 100%; Rabbit: 86%.

TESPA1 is a critical component of the TCR signalosome and is essential for T cell selection and maturation through the regulation of TCR signaling during T cell development. TESPA1 encodes a protein of 458 amino acids and is highly conserved among vertebrate species. TESPA1 expression was tightly regulated during T cell development, with highest expression at the DP stage. TESPA1 deficiency resulted in failure of positive selection and substantially fewer of CD4+ and CD8+ SP cells in the thymus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TESPA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TESPA1
Gene Accession No. A2RU30
Gene Alias 5830405N20Rik; A430001F24Rik; AI450549; HSPC257; KIAA0748; LOW QUALITY PROTEIN: protein TESPA1; protein TESPA1; TESPA1; thymocyte expressed; thymocyte expressed, positive selection associated 1; thymocyte-expressed positive selection-associated protein 1
Gene Symbols TESPA1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TESPA1 (aa 394-443).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9840
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.