Learn More
Fisherbrand™ Test Paper
Shop All Fisherbrand Products
Click to view available options
Type:
Cobalt Chloride Test Paper
Lead Acetate Test Paper
Litmus Blue Test Strip
Litmus Neutral Test Strip
Litmus Red Test Strip
Methyl Violet Test Strip
Moisture Detection Test Strip
PTC Taste Test Paper
Description
- PTC (pheylthiocarbamide) taste test paper test for the genetically controlled ability to taste this substance
- This will allow for the development of a Pedigree for an individual
- PTC can taste bland, bitter or even vile depending upon how sensitive your taste buds are to this chemical
- The bitter taste detected by some individuals, and its intensity, depends upon a homozygous dominant or heterozygous condition in the individuals tested
- PTC Taste Test Paper is perfectly safe to use
- Phenylthiocarbamide (PTC) is present at only 3-5 micrograms per strip
- At this level, the compound is negligible and harmless
- Cobalt Chloride test paper detects the presence of moisture
- This test paper is useful in difficult to reach places and recesses
- Cobalt chloride test paper is responsive to 50% relative humidity and above, and any direct measure of an aqueous solution
- Phenolphthalein test paper is sensitive to Ammonia Vapors, and thus is used to detect Ammonia leaks, for example, in refrigerators or air conditioners
- Phenolphthalein detects Ammonia in solution from 3 - 5ppm
- The test strip will turn a pinkish color if Ammonia is detected
Specifications
Specifications
| For Use With (Application) | For all laboratory moisture tests |
| Quantity | Pk. of 12 Vials; 100 Strips/Vial |
| Type | Methyl Violet Test Strip |