Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Test Paper
SCI_ED

Catalog No. S67170 Shop All Fisherbrand Products
Click to view available options
Type:
Cobalt Chloride Test Paper
Lead Acetate Test Paper
Litmus Blue Test Strip
Litmus Neutral Test Strip
Litmus Red Test Strip
Methyl Violet Test Strip
Moisture Detection Test Strip
PTC Taste Test Paper
This item is not returnable. View return policy
This item is not returnable. View return policy

Use these test papers for a variety of applications.

  • A foolproof pH test to determine if a solution is acidic
  • The Blue Litmus test paper does not come with a color chart as it cannot provide you with a defined pH value
  • The test strip will turn red if dipped into an acidic solution (pH <7)
  • It will remain unchanged in an alkaline solution
  • We have tested the blue litmus with multiple pH buffers
  • The blue paper is blue at pH 8
  • At pH 7, the sample is maybe just a little red
  • The real change to red seems to start with pH 6
  • 5 and lower
  • Neutral Litmus test paper tests for Neutrality of a solution, or its acidity or alkalinity
  • The test strip will turn red if dipped into an acidic solution (pH <7) or blue if dipped into an alkaline solution (pH >7)
  • Red Litmus test paper is a foolproof pH test to determine if a solution is alkaline
  • The test strip will turn blue if dipped into an alkaline solution (pH >7)
  • Color chart not included

Specifications

Certifications/Compliance NFPA
pH Range 0 to 7
For Use With (Application) For all laboratory moisture tests
Quantity Pk. of 12 Vials; 100 Strips/Vial
Type Litmus Blue Test Strip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.