Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QuidelOrtho Test Reagents
Click to view available options
Quantity:
300 Tests
600 Tests
Type:
Amphetamines
Anti-Streptolysin
Apolipoprotein A1
Apolipoprotein B
Barbiturates
Benzodiazepines
Caffeine
Cannabinoids
Cocaine Metabolite
Complement C3
Complement C4
Gentamicin
Haptoglobin
High Sensitivity C-Reactive Protein
IgA
IgG
IgM
Methadone
Microalbumin
Opiates
Prealbumin
Rheumatoid Factor
Tobramycin
Transferrin
Valproic Acid
Vancomycin
alpha-1-antitrypsin
Specifications
Specifications
Certifications/Compliance | Class 7, FDA |
Type | Valproic Acid |
For Use With (Application) | Therapeutic and abused drug testing, specific Proteins |
Quantity | 300 Tests |
Form | Liquid Ready-to-Use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction