Learn More
Eisco™ Test Strips 7-10 Range - Narrow-Range pH Testing Kit with Broad Color Spectrum, 50 Strips
Shop All Eisco Products
Description
- Accurate pH testing: Measures pH levels from 7 to 10 in 0.5 increments, ideal for educational labs and scientific research.
- Versatile uses: Perfect for environmental water testing, ensuring safety and compliance in public and private water supplies.
- Narrow-Range focus: Designed for precise pH measurements, crucial for maintaining proper conditions in instances such as hydroponics and aquariums.
- Easy color comparison: Each strip features a broad color spectrum, simplifying the process of matching against pH levels in gardening.
- Quick results: Obtain immediate and reliable feedback, essential for routine checks in spa and pool maintenance.
- Our pH Test Strips in the 7-10 range are perfect for accurately measuring neutral to mildly alkaline solutions.
- Each package contains 100 strips designed for high sensitivity and precision in critical pH testing scenarios.
- Easy to use, providing quick visual feedback with a detailed color spectrum on each strip that helps distinguish pH levels in 0.5 increments.
- Ideal for a variety of applications, including educational laboratories, gardening pH testing, and other professional environments where pH monitoring is essential.
- These strips are a practical tool for routine checks, ensuring your solutions maintain the desired pH levels for optimal results.
- This product is intended for testing and monitoring purposes only and is not designed for medical use.
Specifications
Specifications
| pH Range | 7 to 10 |
| Type | pH Test Strips 7-10 Range |