Learn More
Testosterone Antibody (14P2C8), Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP178562
Catalog No. NBP178562
Only null left
Add to Cart
Description
Description
Testosterone Monoclonal specifically detects Testosterone in Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Radioimmunoassay.
Specifications
Specifications
Testosterone
Monoclonal
1 mg/mL
ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000
Testosterone-3-CMO-BSA
100 μg
Cellular Signaling, Developmental Biology
Chemical.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
14P2C8
Unconjugated
Mouse
Ion exchange chromatography
RUO
Primary
Other
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only