Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Testosterone Antibody (3T16), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250332AF350
Description
Testosterone Monoclonal specifically detects Testosterone in Non-species specific samples. It is validated for ELISA.Specifications
Testosterone | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein A purified | |
Cellular Signaling, Developmental Biology | |
Non-species specific | |
IgG1 |
ELISA | |
3T16 | |
ELISA | |
17b-Hydroxyandrost-4-ene-3-one, 17b-Testosterone | |
Testosterone-3-(O-carboxymethyl)oxime coupled to bovine serum albumin | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction