Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Testosterone Antibody (4E1G2), Alexa Fluor™ 700 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311603AF700
Description
Testosterone Monoclonal antibody specifically detects Testosterone in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Testosterone | |
Monoclonal | |
Alexa Fluor 700 | |
17b-Hydroxyandrost-4-ene-3-one, 17b-Testosterone | |
Testosterone 3 CMO conjugated to BSA | |
0.1 mL | |
Cellular Signaling, Developmental Biology | |
Non-species specific | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
4E1G2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction