Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Testosterone Antibody (BGN/06/902), DyLight 550

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062942R
Description
Testosterone Monoclonal specifically detects Testosterone in Non-species specific samples. It is validated for ELISA.Specifications
Testosterone | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Testosterone-11-HS-BSA | |
0.1 mL | |
Cellular Signaling, Developmental Biology | |
This product is specific for Testosterone. It has shown the following reactivities. Testosterone 100% 19 Nortestosterone 15% Dihydrotestosterone <1% Testosterone promotes the development and maintenance of male secondary sex characteristics and is produced primarily in the testes. It is also produced in small amounts by the adrenal glands, ovaries and placenta. Synthetic derivatives of testosterone are used therapeutically in the treatment of breast cancer and for hormone replacement therapy. | |
Store at 4C in the dark. | |
IgG3 |
ELISA | |
BGN/06/902 | |
ELISA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction