Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Testosterone Monoclonal Antibody (3G7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549658
Description
Antibody tested in indirect ELISA.
Testosterone is a steroid hormone from the androgen group and is found in mammals, reptiles, birds, and other vertebrates. In mammals, testosterone is primarily secreted in the testes of males and the ovaries of females, although small amounts are also secreted by the adrenal glands. It is the principal male sex hormone and an anabolic steroid.
Specifications
Testosterone | |
Monoclonal | |
4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Chemical | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
3G7 | |
Unconjugated | |
Androderm; C19H28O2; Mertestate; Sustanon; Testoderm; Testosteron | |
Conjugate of T hapten and KLH. | |
200 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction