Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Testosterone Monoclonal Antibody (4E1G2)
GREENER_CHOICE

Catalog No. PIMA183483
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183483 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183483 Supplier Invitrogen™ Supplier No. MA183483
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product has an Affinity of 2.3x10^10 L/mole. Mouse anti Testosterone (4E1G2) recognizes Testosterone, which promotes the development and maintenance of male secondary sex characteristics and is produced primarily in the testes.

Testosterone is a steroid hormone from the androgen group and is found in mammals, reptiles, birds, and other vertebrates. In mammals, testosterone is primarily secreted in the testes of males and the ovaries of females, although small amounts are also secreted by the adrenal glands. It is the principal male sex hormone and an anabolic steroid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Testosterone
Applications ELISA
Classification Monoclonal
Clone 4E1G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias Androderm; C19H28O2; Mertestate; Sustanon; Testoderm; Testosteron
Host Species Mouse
Immunogen Testosterone conjugated to BSA.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.