Learn More
Invitrogen™ Testosterone Polyclonal Antibody

Description
Working Concentration: 10 μg/mL. Sensitivity: 10 ng/mL Testosterone produces 83% inhibition in a competitive ELISA employing Testosterone polyclonal antibody.
Specifications
Specifications
| Antigen | Testosterone |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 6.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.3 |
| Gene Alias | Androderm; C19H28O2; Mertestate; Sustanon; Testoderm; Testosteron |
| Host Species | Sheep |
| Immunogen | Testosterone(17)-BSA. |
| Purification Method | Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.