Learn More
Invitrogen™ Testosterone Recombinant Rabbit Monoclonal Antibody (RM435)

Description
Testosterone Recombinant Monoclonal Antibody for ELISA
Specifications
Specifications
| Antigen | Testosterone |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Clone | RM435 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide |
| Gene Alias | Androderm; C19H28O2; Mertestate; Sustanon; Testoderm; Testosteron |
| Host Species | Rabbit |
| Immunogen | Testosterone-KLH. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.