Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Tet Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144574
Description
Tet Polyclonal Antibody for Western Blot, ELISA
Dioxygenase that specifically demethylates DNA methylated on the 6th position of adenine (N(6)-methyladenosine) DNA (PubMed:25936838, PubMed:30078725). N(6)-methyladenosine (m6A) DNA is present at a relatively high level at the very earliest embryonic stages but at low levels at the late embryonic stages and may act as a regulator of gene expression (PubMed:25936838). Promotes differentiation of early germ cells in ovary (PubMed:25936838). Contributes to neuronal morphology, development, and function in the brain (PubMed:30078725). By interacting with histone modifier wds, binds to a specific set of genes, modulates intragenic (N(6)-methyladenosine) DNA levels and thereby maintains transcriptional activation (PubMed:30078725). Also able to catalyze the conversion of the modified genomic base 5-methylcytosine (5mC) into 5-hydroxymethylcytosine (5hmC) (PubMed:25936838).
Specifications
Tet | |
Polyclonal | |
Unconjugated | |
Tet | |
DMAD | |
Rabbit | |
Affinity chromatography | |
RUO | |
38347 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2.808 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
M9NEY8 | |
Tet | |
Recombinant Fruit fly Tet protein (2494-2767aa). | |
100 μL | |
Primary | |
Drosophila | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction