Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tet Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144404
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144404 Supplier Invitrogen™ Supplier No. PA5144404
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tet Polyclonal Antibody for Western Blot, ELISA

Dioxygenase that specifically demethylates DNA methylated on the 6th position of adenine (N(6)-methyladenosine) DNA (PubMed:25936838, PubMed:30078725). N(6)-methyladenosine (m6A) DNA is present at a relatively high level at the very earliest embryonic stages but at low levels at the late embryonic stages and may act as a regulator of gene expression (PubMed:25936838). Promotes differentiation of early germ cells in ovary (PubMed:25936838). Contributes to neuronal morphology, development, and function in the brain (PubMed:30078725). By interacting with histone modifier wds, binds to a specific set of genes, modulates intragenic (N(6)-methyladenosine) DNA levels and thereby maintains transcriptional activation (PubMed:30078725). Also able to catalyze the conversion of the modified genomic base 5-methylcytosine (5mC) into 5-hydroxymethylcytosine (5hmC) (PubMed:25936838).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tet
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.475 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Tet
Gene Accession No. M9NEY8
Gene Alias DMAD
Gene Symbols Tet
Host Species Rabbit
Immunogen Recombinant Drosophila melanogaster (Fruit fly) DMAD protein (2494-2767aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 38347
Target Species Drosophila
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.