Learn More
Invitrogen™ Tet Repressor Polyclonal Antibody

Description
PA1-4501 detects Tet repressor in bacteria samples. PA1-4501 has been successfully used in Western blot and ELISA procedures. The PA1-4501 immunogen is Residues 31-49 [AQKLGVEQPTLYWHVKNKR] of the 22 kDa Tet repressor protein. Maintain at 2-8ºC of 3 months or at -20ºC for longer periods. Avoid repeated freeze/thaw cycles.
Specifications
Specifications
| Antigen | Tet Repressor |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | pSC101 tetR; TetR |
| Host Species | Rabbit |
| Immunogen | Residues 31-49 [AQKLGVEQPTLYWHVKNKR] of the 22 kDa Tet repressor protein. |
| Purification Method | Antigen Affinity Chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.