Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TET1 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178966F
Description
TET1 Polyclonal specifically detects TET1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC/IF (Negative).Specifications
TET1 | |
Polyclonal | |
Immunohistochemistry 1:400, Immunohistochemistry-Paraffin 1:400, Western Blot (Negative), ICC/IF (Negative) | |
Q8NFU7 | |
TET1 | |
A synthetic peptide made to an internal portion of the human TET1 protein (between residues 1700 to 1800). [UniProt Q8NFU7] | |
0.1 mL | |
Primary | |
Mouse | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
FITC | |
PBS with 0.05% Sodium Azide | |
bA119F7.1, CXXC finger 6, CXXC6, CXXC6FLJ10839, FLJ41442, KIAA1676CXXC-type zinc finger protein 6, LCX, LCXEC 1.14.11.n2, Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, Ten-eleven translocation 1 gene protein, ten-eleven translocation-1, tet oncogene 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
80312 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction