Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TET1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586010 Supplier Invitrogen™ Supplier No. PA586010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TET1 Polyclonal Antibody for Flow, ICC/IF, IHC (P)

Dioxygenase that specifically binds methylcytosine (5mC), a minor base in mammalian DNA found in repetitive DNA elements that is crucial for retrotransposon silencing and mammalian development. Catalyzes the conversion of methylcytosine (5mC) to 5-hydroxymethylcytosine (hmC). The clear function of 5-hydroxymethylcytosine (hmC) is still unclear but it may influence chromatin structure and recruit specific factors or may constitute an intermediate component in cytosine demethylation. 5-hydroxymethylcytosine (hmC) is present in ES cells and is enriched in the brain, especially in Purkinje neurons. May play a role in the fetal development of heart, lung and brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TET1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene TET1
Gene Accession No. Q3URK3, Q8NFU7
Gene Alias 2510010B09Rik; AA517754; bA119F7.1; BB001228; CXXC finger 6; CXXC zinc finger 6; CXXC6; CXXC-type zinc finger protein 6; D10Ertd17e; KIAA1676; LCX; leukemia-associated protein with a CXXC domain; Methylcytosine dioxygenase TET1; mKIAA1676; MLL-TET1; RP11-119F7.1; Ten-eleven translocation 1 gene protein; ten-eleven translocation 1 gene protein homolog; ten-eleven translocation-1; tet methylcytosine dioxygenase 1; tet oncogene 1; Tet1; TET1 splice variant VP_DE4; TET1 splice variant VP_DE456; TET1-MLL
Gene Symbols TET1
Host Species Rabbit
Immunogen Synthetic peptide within Human Tet1 aa 851-900.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309902, 52463, 80312
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.