Learn More
Invitrogen™ TET1 Polyclonal Antibody

Description
Immunogen sequence: VPPNPIATFN APSKWPEPQS TVSYGLAVQG AIQILPLGSG HTPQSSSNSE KNSLPPVMAI SNVENEKQVH ISFLPANTQG FPLAPERGLF HASLGIAQLS QAGPSKSDRG SSQVSVTSTV HVVNTTVVTM PVPMVSTSSS SYTT Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 47%.
Specifications
Specifications
| Antigen | TET1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TET1 |
| Gene Accession No. | Q8NFU7 |
| Gene Alias | 2510010B09Rik; AA517754; bA119F7.1; BB001228; CXXC finger 6; CXXC zinc finger 6; CXXC6; CXXC-type zinc finger protein 6; D10Ertd17e; KIAA1676; LCX; leukemia-associated protein with a CXXC domain; Methylcytosine dioxygenase TET1; mKIAA1676; MLL-TET1; RP11-119F7.1; Ten-eleven translocation 1 gene protein; ten-eleven translocation 1 gene protein homolog; ten-eleven translocation-1; tet methylcytosine dioxygenase 1; tet oncogene 1; Tet1; TET1 splice variant VP_DE4; TET1 splice variant VP_DE456; TET1-MLL |
| Gene Symbols | TET1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.