Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TET1, Rat, Unlabeled, Clone: 5D6,

Catalog No. MABE1144MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE1144MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1144MI Supplier MilliporeSigma™ Supplier No. MABE1144
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Methylcytosine dioxygenase TET1 (EC 1.14.11.n2; UniProt Q3URK3; also known as CXXC-type zinc finger protein 6, Ten-eleven translocation 1 gene protein homolog) is encoded by the Tet1 (also know as Cxxc6, Kiaa1676) gene (Gene ID 52463) in murine species. TET (ten-eleven translocation) proteins (TET1, TET2, and TET3) oxidize 5-methylcytosine to 5-hydroxymethylcytosine, 5-formylcytosine, and 5-carboxylcytosine, providing a means for active epigenetic DNA demethylation in mammals. TET proteins also modify genomic thymine residues to 5-hydroxyuracil. TET proteins depend on Fe(II) and 2-oxoglutarate as cofactors for activity, and 2-oxoglutarate is known to inhibit their activity, while ascorbate is shown to stimulate TET-mediated cytosine oxidation. In addition, post-translational modifications (PTMs), such as phosphorylation and O-GlcNAcylation, also play a role in regulating the activity of TET proteins. TET1 and TET2 are found highly expressed in mouse embryonic stem cells (mESCs) and TET3 is known to be upregulated in oocytes and oxidize the silenced paternal pronuclear DNA. High levels of TET proteins and genomic 5-hydroxymethylcytosine in neuronal tissues are also reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TET1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5D6
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Tet1, Cxxc6, Kiaa1676
Gene Accession No. Q3URK3
Host Species Rat
Immunogen His-tagged recombinant fragment from the region between DNA-binding and 2-oxoglutarate-binding domains of mouse TET1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_081660
Test Specificity Clone 5D6 detected exogenously expressed TET1, but not TET2 or TET3, GST fusion protein (Bauer, C., et al. (2015). J. Biol. Chem. 290(8):4801-4812).
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.