Learn More
TET2 Antibody (442), Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP243579
Catalog No. NBP243579
Only null left
Add to Cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Immunoprecipitation
TET2 Monoclonal specifically detects TET2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
Specifications
TET2
Monoclonal
Unconjugated
PBS with No Preservative
TET2
Recombinant protein encompassing a sequence within the N-terminus region of human TET2. The exact sequence is proprietary.
0.1 mL
Primary
Human
Purified
Western Blot, Immunoprecipitation
442
Western Blot 1:1000-1:10000, Immunoprecipitation 1:100-1:500
FLJ20032, MGC125715, tet methylcytosine dioxygenase 2
Mouse
Protein G purified
RUO
54790
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only