Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TET2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232104AF647
Description
TET2 Polyclonal antibody specifically detects TET2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
TET2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
FLJ20032, MGC125715, tet methylcytosine dioxygenase 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
54790 | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TET2 | |
The immunogen this antibody was made to, maps to a region between residue 100 and 150 of human Tet Oncogene Family Member 2 using the numbering given in entry NP_001120680.1 (GeneID 54790). | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction