Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TET3 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298069JF549
Description
TET3 Polyclonal specifically detects TET3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TET3 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
EC 1.14.11.n2, hCG_40738, KIAA0401, methylcytosine dioxygenase TET3, MGC22014, probable methylcytosine dioxygenase TET3, tet oncogene family member 3 | |
Produced in rabbits immunized with E. coli-derived Human TET3 fragment. | |
0.1 mL | |
Epigenetics | |
200424 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction