Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TET3 Monoclonal Antibody (GT334)
GREENER_CHOICE

Catalog No. PIMA527863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527863 Supplier Invitrogen™ Supplier No. MA527863
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: flag-human TET3-transfected 293T cells Predicted Reactivity: Mouse (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TET3, a member of the ten-eleven-translocation (TET) family of genes, is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine and is most abundantly expressed in hematopoietic cells. Unlike the related TET2 protein, mutations in TET3 have not been observed in any myeloid malignancies. TET3 has been shown to be involved in the demethylation of zygotic DNA before the first mitosis and has been suggested to be involved in the epigenetic reprogramming of the zygotic paternal DNA following natural fertilization and may also contribute to somatic cell nuclear reprogramming during animal cloning.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TET3
Applications Western Blot
Classification Monoclonal
Clone GT334
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TET3
Gene Accession No. O43151
Gene Alias B430006D22Rik; BC037432; D230004J03Rik; hCG_40738; KIAA0401; Methylcytosine dioxygenase TET3; MGC22014; probable methylcytosine dioxygenase TET3; putative methylcytosine dioxygenase; Tet methylcytosine deoxygenase 3 isoform; tet methylcytosine dioxygenase 3; tet oncogene family member 3; Tet3
Gene Symbols TET3
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human TET3. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200424
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.