Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TET3, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABE290 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE290 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE290 Supplier MilliporeSigma™ Supplier No. ABE290
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MABE1144/1132/1133 Methylcytosine dioxygenase TET3 (EC 1.14.11.n2; UniProt O43151) is encoded by the TET3 (also known as KIAA0401) gene (Gene ID 200424) in human. TET (ten-eleven translocation) proteins (TET1, TET2, and TET3) oxidize 5-methylcytosine (5mC) to 5-hydroxymethylcytosine (5hmC), 5-formylcytosine (5fC), and 5-carboxylcytosine (5caC), providing a means for active epigenetic DNA demethylation in mammals. TET proteins also modify genomic thymine residues to 5-hydroxyuracil. TET proteins depend on Fe(II) and 2-oxoglutarate as cofactors for activity, and 2-oxoglutarate is known to inhibit their activity, while ascorbate is shown to stimulate TET-mediated cytosine oxidation. In addition, post-translational modifications (PTMs), such as phosphorylation and O-GlcNAcylation, also play a role in regulating the activity of TET proteins. TET1 and TET2 are found highly expressed in mouse embryonic stem cells (mESCs) and TET3 is known to be upregulated in oocytes and oxidize the silenced paternal pronuclear DNA. High levels of TET proteins and genomic 5-hydroxymethylcytosine in neuronal tissues are also reported. Three TET3 spliced isoforms/variants have been reported, the long or full-length isoform Tet3FL and the short isoform Tet3s are upregulated during embryonic stem cell (ESC) differentiation toward the neuronal lineage, while Tet3o is oocyte-specific and not detected in ESCs or in any other cell types or adult mouse tissues tested, indicating a role of Tet3o in paternal genome oxidation in mouse zygotes. Tet3FL and Tet3o contain the entire Tet3s sequence with additional (but different) N-terminal sequence. Only Tet3FL contains an N-terminal CXXC domain that binds to unmethylated CpGs and exhibits highest affinity toward 5caC. Tet3FL is substantially less active than Tet3o and Tet3s, suggesting that the CXXC domain restricts Tet3FL 5mC oxidation capacity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TET3
Applications ChIP Assay, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene TET3, KIAA0401
Gene Accession No. O43151
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corrsponding to an internal sequence within the N-terminal half of human TET3.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001274420
Test Specificity Immunogen sequence is present in TET3 isoforms Tet3FL, Tet3s, and Tet3o spliced isoforms reported by Jin, S.G., et al. (2016). Cell Rep. 14(3):493-505), as well as all additional human and mouse TET3 spliced isoforms reported by UniProt (O43151, Q8BG87). Note that UniProt isoform 1 (O43151-1/human and Q8BG87-1/mouse) corresponds to isoform Tet3s, not Tet3FL (NM_001287491.1/human and JX946278.1/mouse).
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt. Note: It is recommended to store the product undiluted at 2°-8°C and protected from prolonged exposure to light. Do not freeze.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.