Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TetR Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600234AF532
Description
TetR Polyclonal specifically detects TetR in Non-species specific samples. It is validated for Western Blot, ELISA.Specifications
TetR | |
Polyclonal | |
Western Blot, ELISA | |
pSC101 tetR, Tet Repressor, Tet Repressor Protein, TetR, tetracycline repressor protein | |
A synthetic peptide made to an internal portion of Escherichia coli TetR protein (between residues 25-75) [UniProt P04483] | |
0.1 mL | |
Primary | |
Non-species specific | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Rabbit | |
Affinity Purified | |
Transcription Factors and Regulators | |
4924774.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction