Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TetR Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178424AF750
Description
TetR Polyclonal specifically detects TetR in Non-species specific samples. It is validated for Western Blot.Specifications
TetR | |
Polyclonal | |
Western Blot | |
pSC101 tetR, Tet Repressor, Tet Repressor Protein, TetR, tetracycline repressor protein | |
A synthetic peptide made to an N-terminal portion of Escherichia coli TetR protein (between residues 1-50) [UniProt P04483] | |
Primary | |
Non-species specific | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
Rabbit | |
0.1 mL | |
4924774 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction