Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TetR Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB600234
Description
TetR Polyclonal specifically detects TetR in Non-species specific samples. It is validated for Western Blot, ELISA.Specifications
| TetR | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.02% Sodium Azide | |
| pSC101 tetR, Tet Repressor, Tet Repressor Protein, TetR, tetracycline repressor protein | |
| A synthetic peptide made to an internal portion of Escherichia coli TetR protein (between residues 25-75) [UniProt P04483] | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 4924774 | |
| Non-species specific | |
| IgG |
| Western Blot, ELISA | |
| 1.0 mg/mL | |
| Western Blot 1:100, ELISA 1:100-1:2000 | |
| P04483 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Non-species specific. This antibody is used to monitor the regulation of gene expression in mammalian cells. The TetR antibody is not reactive to human or other mammalian proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction