Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TetR Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313057
This item is not returnable.
View return policy
Description
TetR Polyclonal antibody specifically detects TetR in All species samples. It is validated for Western Blot, ELISASpecifications
| TetR | |
| Polyclonal | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| All species | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| pSC101 tetR, Tet Repressor, Tet Repressor Protein, TetR, tetracycline repressor protein | |
| Full length TetR recombinant protein. | |
| 100 μg | |
| Transcription Factors and Regulators | |
| 4924774 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction