Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetra-n-butylammonium fluoride, 1M soln. in THF
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetra-n-butylammonium fluoride, 1M soln. in THF is used as a reactant for the preparation of triple monoamine reuptake inhibitors as a new generation of antidepressants, oligoribonucleotides with phosphonate-modified linkages, conjugated dienoic acid esters using Suzuki coupling reactions and polyacetylenic glucosides. It plays an important role in aryl alkyl alcohols through Nozaki-Hiyama allylation catalyzed by chiral bipyridyldiol ligands and chromium trichloride. It is also used as a catalyst for the silylation reactions. In addition, it is used as a source of fluoride ion in organic solvents as well as for the deprotection of silyl ether groups.
Solubility
Miscible with terahydrofuran, acetonitrile, dimethyl sulfoxide and organic solvents.
Notes
Store in a cool place. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and oxygen.
Tetra-n-butylammonium fluoride, 1M soln. in THF is used as a reactant for the preparation of triple monoamine reuptake inhibitors as a new generation of antidepressants, oligoribonucleotides with phosphonate-modified linkages, conjugated dienoic acid esters using Suzuki coupling reactions and polyacetylenic glucosides. It plays an important role in aryl alkyl alcohols through Nozaki-Hiyama allylation catalyzed by chiral bipyridyldiol ligands and chromium trichloride. It is also used as a catalyst for the silylation reactions. In addition, it is used as a source of fluoride ion in organic solvents as well as for the deprotection of silyl ether groups.
Solubility
Miscible with terahydrofuran, acetonitrile, dimethyl sulfoxide and organic solvents.
Notes
Store in a cool place. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and oxygen.
Specifications
Specifications
| Density | 0.903 |
| Flash Point | −17°C (1°F) |
| Linear Formula | [CH3(CH2)3]4NF |
| Quantity | 25 mL |
| UN Number | UN2924 |
| Beilstein | 3570522 |
| Sensitivity | Hygroscopic |
| Solubility Information | Miscible with terahydrofuran,acetonitrile,dimethyl sulfoxide and organic solvents. |
| Formula Weight | 261.47 |
| Concentration or Composition (by Analyte or Components) | 1M soln. in THF |
| Show More |
Safety and Handling
| Hazard Category | H225-H314-H335-H336-H351-H373 |
| Hazard Statement | GHS H Statement H225-H314-H351-H302-H335 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Suspected of causing cancer. Harmful if swallowed. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| EINECSNumber | 207-057-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction