Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butylammonium hexafluorophosphate, 98%

Catalog No. AAA1719622
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 3109-63-5 | C16H36F6NP | 387.44 g/mol

Tetra-n-butylammonium hexafluorophosphate is used as a supporting electrolyte in nonaqueous electrochemistry. It is chemically inert, thereby allowing the usage as an inert electrolyte with potential range. It enhances the performance of single-layer organic light emitting diodes (OLEDs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium hexafluorophosphate is used as a supporting electrolyte in nonaqueous electrochemistry. It is chemically inert, thereby allowing the usage as an inert electrolyte with potential range. It enhances the performance of single-layer organic light emitting diodes (OLEDs).

Solubility
Soluble in acetone, acetonitrile and tetrhydrofuran.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3109-63-5
Molecular Formula C16H36F6NP
Molecular Weight (g/mol) 387.44
MDL Number MFCD00011748
InChI Key BKBKEFQIOUYLBC-UHFFFAOYSA-N
Synonym tetrabutylammonium hexafluorophosphate, tetra-n-butylammonium hexafluorophosphate, tetrabutylammonium hexafluorophosphate v, 1-butanaminium, n,n,n-tributyl-, hexafluorophosphate 1-, 1-butanaminium, n,n,n-tributyl-, hexafluorophosphate 1-1:1, acmc-1ahjo, n,n,n-tributyl-1-butanaminium hexafluorophosphate, ksc225q6r, tetrabutylammoniumhexafluorophosphate, tetrabutylazanium hexafluorophosphate
PubChem CID 165075
SMILES F[P-](F)(F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 242°C to 248°C
Linear Formula [CH3(CH2)3]4NPF6
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in acetone,acetonitrile and tetrhydrofuran.
IUPAC Name hexafluoro-λ⁵-phosphanuide; tetrabutylazanium
Formula Weight 387.44
Percent Purity 98%
Chemical Name or Material Tetra-n-butylammonium hexafluorophosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-472-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.