Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butylammonium nitrate, 98%

Catalog No. AA4334222
Click to view available options
Quantity:
5 g
25 g
100 g
500 g

Catalog No. AA4334222

Supplier: Thermo Scientific Chemicals 04334222

Only null left
Add to Cart

CAS: 1941-27-1 | C16H36N2O3 | 304.475 g/mol

Tetra-n-butylammonium nitrate is used in coatings, printing ink, rubber, glass, leather and cosmetics. It serves as a supporting electrolyte in liquid ammonia. It is also used in the preparation of nitrates from triflates. It is employed in the inversion of configuration of alcohols through tosylates. It is also a source of NO3- used in nucleophilic substitution reactions. Further, it is used in the conversion of halides into ketones. In addition to this, it is used as a source of nitronium ions for the nitration of olefins, amides, ribonucleosides, glycols and aromatic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1941-27-1
Molecular Formula C16H36N2O3
Molecular Weight (g/mol) 304.475
MDL Number MFCD00043202
InChI Key QHOKENWFMZXSEU-UHFFFAOYSA-N
Synonym tetrabutylammonium nitrate, tetra-n-butylammonium nitrate, 1-butanaminium, n,n,n-tributyl-, nitrate, tetrabutylazanium nitrate, ammonium, tetrabutyl-, nitrate, n-n-n-tributyl-1-butanaminium nitrate, 1-butanaminium, n,n,n-tributyl-, nitrate 1:1, tetrabutylammonium ion nitrate, tetrabutylammoniumnitrate, acmc-209exx
PubChem CID 16027
IUPAC Name tetrabutylazanium;nitrate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[N+](=O)([O-])[O-]

Specifications

Melting Point 116°C to 118°C
Odor Weak
Quantity 100 g
UN Number UN1479
Sensitivity Hygroscopic
Solubility Information Soluble in dichloromethane,acetonitrile,benzene and toluene.
Formula Weight 304.48
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tetra-n-butylammonium nitrate
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P220-P221-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P372-P375-P380-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 217-726-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.