Learn More
Tetra-n-butylammonium nitrate, 98%
Description
Tetra-n-butylammonium nitrate is used in coatings, printing ink, rubber, glass, leather and cosmetics. It serves as a supporting electrolyte in liquid ammonia. It is also used in the preparation of nitrates from triflates. It is employed in the inversion of configuration of alcohols through tosylates. It is also a source of NO3- used in nucleophilic substitution reactions. Further, it is used in the conversion of halides into ketones. In addition to this, it is used as a source of nitronium ions for the nitration of olefins, amides, ribonucleosides, glycols and aromatic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 116°C to 118°C |
| Odor | Weak |
| Quantity | 5 g |
| UN Number | UN1479 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in dichloromethane,acetonitrile,benzene and toluene. |
| Formula Weight | 304.48 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | Tetra-n-butylammonium nitrate |
Safety and Handling
| Hazard Category | H272-H315-H319-H335 |
| Hazard Statement | GHS H Statement H272-H315-H319-H335 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P220-P221-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P372-P375-P380-P501c |
| DOTInformation | Hazard Class: 5.1; Packaging Group: II |
| EINECSNumber | 217-726-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.