Alfa Aesar™ Tetra-n-butylammonium perchlorate, electrochemical grade

Suitable for use as a polarographic supporting electrolyte

Manufacturer: Alfa Aesar™ 4399909

Quantity 10g

 View more versions of this product

Catalog No. AA4399909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1923-70-2
    Molecular Formula C16H36ClNO4
    Linear Formula [CH3(CH2)3]4NClO4
    Formula Weight 341.92
    MDL Number MFCD00038722
    UN Number UN1481
    Chemical Name or Material Tetra-n-butylammonium perchlorate
    Grade Electrochemical grade

    Specifications

    Formula Weight 341.92
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Crystalline
    Health Hazard 1 Danger
    Linear Formula [CH3(CH2)3]4NClO4
    Solubility Information Soluble in ethanol; Very slightly soluble in water
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
    Molecular Formula C16H36ClNO4
    CAS 1923-70-2
    UN Number UN1481
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 210° to 217°C
    Quantity 10g
    Sensitivity Hygroscopic
    MDL Number MFCD00038722
    Color White
    Chemical Name or Material Tetra-n-butylammonium perchlorate
    Grade Electrochemical grade

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit