Learn More
Tetra-n-butylammonium perchlorate, electrochemical grade
Description
Suitable for use as a polarographic supporting electrolyteTetra-n-butylammonium perchlorate is used as a supporting electrolyte in polarographic measurements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 210°C to 217°C |
Color | White |
Linear Formula | [CH3(CH2)3]4NClO4 |
Quantity | 50 g |
UN Number | UN1481 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in acetonitrile and ethanol; Very slightly soluble in water. |
Formula Weight | 341.92 |
Grade | Electrochemical |
Physical Form | Crystalline |
Show More |
Safety and Handling
Hazard Category | H272-H315-H319-H335 |
Hazard Statement | GHS H Statement H272-H315-H319-H335 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S. |
EINECSNumber | 217-655-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.