Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-octylammonium bromide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
14866-33-2
C32H68BrN
546.81
MFCD00011863
QBVXKDJEZKEASM-UHFFFAOYSA-M
tetraoctylammonium bromide,tetra-n-octylammonium bromide,tetraoctylazanium bromide,tetraoctyl ammonium bromide,1-octanaminium, n,n,n-trioctyl-, bromide,unii-52qw29owoc,52qw29owoc,tetra n-octyl ammonium bromide,acmc-1bvth,tetra-octylammonium bromide
2734117
tetraoctylazanium;bromide
CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC.[Br-]
14866-33-2
C32H68BrN
546.81
MFCD00011863
QBVXKDJEZKEASM-UHFFFAOYSA-M
tetraoctylammonium bromide,tetra-n-octylammonium bromide,tetraoctylazanium bromide,tetraoctyl ammonium bromide,1-octanaminium, n,n,n-trioctyl-, bromide,unii-52qw29owoc,52qw29owoc,tetra n-octyl ammonium bromide,acmc-1bvth,tetra-octylammonium bromide
2734117
tetraoctylazanium;bromide
CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC.[Br-]
Phase transfer catalyst
Melting Point 95°C to 99°C
Quantity 5g
Packaging Glass bottle
Color Cream-White to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,dichloromethane and chloroform
Formula Weight 546.81
Physical Form Powder or Flakes
Percent Purity 98%
Chemical Name or Material Tetra-n-octylammonium bromide
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 238-936-9